Learn More
Raybiotech Inc Human Lymphotoxin beta R/TNF RIII/TNFRSF3 ELISA Kit
Supplier:  Raybiotech Inc ELHLTBR1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Lymphotoxin beta R.
Catalog No. 50-194-9513