Learn More
Raybiotech Inc Human Proprotein Convertase 9/PCSK9 ELISA Kit
Supplier:  Raybiotech Inc ELHPCSK91
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Proprotein Convertase 9.
Catalog No. 50-194-9628