Learn More
Raybiotech Inc Human Pancreatic Polypeptide/PP ELISA Kit
Supplier:  Raybiotech Inc ELHPPY1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Pancreatic Polypeptide/PP.
Catalog No. 50-194-9658