Learn More
Raybiotech Inc Human Calprotectin L1/S100-A8/A9 Complex ELISA Kit
Supplier:  Raybiotech Inc ELHS100A891
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 35 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Calprotectin L1.
Catalog No. 50-194-9696