Learn More
Raybiotech Inc Human TIMP-1 ELISA Kit
Supplier:  Raybiotech Inc ELHTIMP1CL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 30 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human TIMP-1.
Catalog No. 50-194-9784