Learn More
Raybiotech Inc Human Tripeptidyl-peptidase I/TPP1 ELISA Kit
Supplier:  Raybiotech Inc ELHTPP11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Tripeptidyl-peptidase I.
Catalog No. 50-194-9800