Learn More
Raybiotech Inc Rhesus Monkey Cathepsin S ELISA Kit
Supplier:  Raybiotech Inc ELKCATHEPSINS1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 28 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rhesus Macaque Cathepsin S (CTSS)
Catalog No. 50-194-9870