Learn More
Raybiotech Inc Rhesus Macaque MCP-1/CCL2 ELISA Kit
Supplier:  Raybiotech Inc ELKMCP11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 8 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rhesus Macaque MCP-1.
Catalog No. 50-194-9891