Learn More
Raybiotech Inc Mouse Coagulation Factor III (Tissue Factor/Thromboplastin) ELISA Kit
Supplier:  Raybiotech Inc ELMCOAGFIII1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Mouse Coagulation Factor III (F3 / Tissue Factor / Thromboplastin)
Catalog No. 50-194-9951