Learn More
Raybiotech Inc Mouse Follistatin-like 1/FSTL1 ELISA Kit
Supplier:  Raybiotech Inc ELMFSTL11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.43 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse Follistatin-like 1.
Catalog No. 50-194-9992