Learn More
Raybiotech Inc Mouse VEGF-A ELISA Kit
Supplier:  Raybiotech Inc ELMVEGFCL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse VEGF.
Catalog No. 50-195-0161