Learn More
Raybiotech Inc Porcine IL-1 alpha/IL-1 F1 ELISA Kit
Supplier:  Raybiotech Inc ELPIL1A1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Porcine IL-1 alpha.
Catalog No. 50-195-0-194