Learn More
Raybiotech Inc Human/Mouse Phospho-4E-BP1 (T36) and Total 4E-BP1 ELISA Kit.
Supplier:  Raybiotech Inc PEL4EBP1T36T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated 4E-BP1 (Thr36) and Total 4E-BP1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human and mouse 4E-BP1 phosphorylated at site Threonine-36 and total 4E-BP1.
Catalog No. 50-195-0507