Learn More
Raybiotech Inc Human, Mouse Phospho-ATF2 (Thr69/71) and Total ATF2 ELISA Kit.
Supplier:  Raybiotech Inc PELATF2T69T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated ATF2 (Thr69/71) and Total ATF2 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse ATF2 phosphorylated at site Threonine-69/71 as well as total ATF2.
Catalog No. 50-195-0520