Learn More
Raybiotech Inc Human/Rat Phospho-ACC1 (S79) and Total ACC1 ELISA Kit.
Supplier:  Raybiotech Inc PELACC1S79T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated ACC1 (Ser79) and Total ACC1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, and rat ACC1 phosphorylated at site Serine-79 and Total ACC1.
Catalog No. 50-195-0511