Learn More
Raybiotech Inc Human Phospho-Caveolin-1 (Tyr14) and Total Caveolin-1 ELISA Kit.
Supplier:  Raybiotech Inc PELCAVEOLIN1Y14T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated Caveolin-1 (Tyr14) and Total Caveolin-1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human Caveolin-1 phosphorylated at site Tyrosine-14 as well as total Caveolin-1.
Catalog No. 50-195-0535