Learn More
Raybiotech Inc Human Phospho-CDK1 (Thr161) and Total CDK1 ELISA Kit.
Supplier:  Raybiotech Inc PELCDK1T161T1
Encompass_Preferred
This assay semi-quantitatively measures CDK1 phosphorylated at Threonine-161 as well as total CDK1 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human CDK1 phosphorylated at site Threonine-161 as well as total CDK1.
Catalog No. 50-195-0537