Learn More
Raybiotech Inc Human Phospho-DDR1 (Tyr792) ELISA Kit.
Supplier:  Raybiotech Inc PELDDR1Y7921
Encompass_Preferred
This assay semi-quantitatively measures DDR1 phosphorylated at Tyrosine-792 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human DDR1 phosphorylated at site Tyrosine-792.
Catalog No. 50-195-0549