Learn More
Raybiotech Inc Human Phospho-DDR1 (Tyr792) and Total DDR1 ELISA Kit.
Supplier:  Raybiotech Inc PELDDR1Y792T1
Encompass_Preferred
This assay semi-quantitatively measures DDR1 phosphorylated at Tyrosine-792 as well as total DDR1 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human DDR1 phosphorylated at site Tyrosine-792 as well as total DDR1.
Catalog No. 50-195-0550