Learn More
Raybiotech Inc Human and Mouse Phosphotyrosine EGFR ELISA Kit.
Supplier:  Raybiotech Inc PELEGFRY1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated EGFR (Tyr1086) and Total EGFR in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-EGFR.
Catalog No. 50-195-0556