Learn More
Raybiotech Inc Human Phospho-Estrogen receptor (Ser118) and Total Estrogen receptor ELISA Kit.
Supplier:  Raybiotech Inc PELERAS118T1
Encompass_Preferred
This assay semi-quantitatively measures Estrogen receptor phosphorylated at Serine-118 as well as total Estrogen receptor in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human Estrogen receptor phosphorylated at site Serine-118 as well as total Estrogen receptor .
Catalog No. 50-195-0578