Learn More
Raybiotech Inc Human, Mouse Phospho-FOXO3 (Ser253) and Total FOXO3 ELISA Kit.
Supplier:  Raybiotech Inc PELFOXO3S253T1
Encompass_Preferred
This assay semi-quantitatively measures FOXO3 phosphorylated at Serine-263 as well as total FOXO3 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse FOXO3 phosphorylated at site Serine-263 as well as total FOXO3.
Catalog No. 50-195-0596