Learn More
Raybiotech Inc Human Phospho-FLT3 (Tyr589) and Total FLT3 ELISA Kit.
Supplier:  Raybiotech Inc PELFLT3Y589T1
Encompass_Preferred
This assay semi-quantitatively measures FLT3 phosphorylated at Tyrosine-589 as well as total FLT3 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human FLT3 phosphorylated at site Tyrosine-589 as well as total FLT3.
Catalog No. 50-195-0594