Learn More
Raybiotech Inc Human, Mouse and Rat Phosphotyrosine FRS2 ELISA Kit.
Supplier:  Raybiotech Inc PELFRS2Y1
Encompass_Preferred
This assay semi-quantitatively measures tyrosine-phosphorylated FRS2 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes human FRS2 phosphotyrosine.
Catalog No. 50-195-0598