Learn More
Raybiotech Inc Human, Mouse, Rat Phospho-HDAC2 (Ser394) and Total HDAC2 ELISA Kit.
Supplier:  Raybiotech Inc PELHDAC2S394T1
Encompass_Preferred
This assay semi-quantitatively measures HDAC2 phosphorylated at Serine-394 as well as total HDAC2 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse, Rat HDAC2 phosphorylated at site Serine-394 as well as total HDAC2.
Catalog No. 50-195-0611