Learn More
Raybiotech Inc Human Phospho-IRF-3 (Ser386) and Total IRF-3 ELISA Kit.
Supplier:  Raybiotech Inc PELIRF3S386T1
Encompass_Preferred
This assay semi-quantitatively measures IRF-3 phosphorylated at Serine-386 as well as total IRF-3 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human IRF-3 phosphorylated at site Serine-386 as well as total IRF-3.
Catalog No. 50-195-0626