Learn More
Raybiotech Inc Human and Mouse Phospho-LSD1 (S112) ELISA Kit.
Supplier:  Raybiotech Inc PELLSD1S1121
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated LSD1 (Ser112) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human and mouse LSD1 phosphorylated at Serine-112.
Catalog No. 50-195-0645