Learn More
Raybiotech Inc Human Phospho-LKB1 (Ser428) and Total LKB1 ELISA Kit.
Supplier:  Raybiotech Inc PELLKB1S428T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated LKB1 (Ser428) and Total LKB1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human LKB1 phosphorylated at site Serine-428 as well as total LKB1.
Catalog No. 50-195-0644