Learn More
Raybiotech Inc Human/Mouse/Rat Phospho-MKK3 (S189) and Total MKK3 ELISA Kit.
Supplier:  Raybiotech Inc PELMKK3S189T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated MKK3 (Ser189) and Total MKK3 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, mouse, and rat MKK3 phosphorylated at site Serine-189 and Total MKK3.
Catalog No. 50-195-0658