Learn More
Raybiotech Inc Human/Mouse/Rat Phospho-MKK3 (S189) ELISA Kit.
Supplier:  Raybiotech Inc PELMKK3S1891
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated MKK3 (Ser189) in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, mouse, and rat PEL phosphorylated at site Serine-189.
Catalog No. 50-195-0657