Learn More
Raybiotech Inc Human Phospho-MSK1 (S376) & Total MSK1 ELISA Kit.
Supplier:  Raybiotech Inc PELMSK1S376T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated MSK1 (Ser376) & Total MSK1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human MSK1 phosphorylated at site Serine-276 and total MSK1
Catalog No. 50-195-0664