Learn More
Raybiotech Inc Human Phospho-PKMYT1 (Thr495) and Total PKMYT1 ELISA Kit.
Supplier:  Raybiotech Inc PELPKMYT1T495T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated PKMYT1 (Thr495) and Total PKMYT1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human PKMYT1 phosphorylated at site Threonine-495 as well as total PKMYT1.
Catalog No. 50-195-0691