Learn More
Raybiotech Inc Human/Mouse/Rat Phospho-PTEN (S380) and Total PTEN ELISA Kit.
Supplier:  Raybiotech Inc PELPTENS380T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated PTEN (Ser380) and Total PTEN in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: Phosphatase and tensin homolog (PTEN) phosphorylated at Serine-380 and Total PTEN
Catalog No. 50-195-0699