Learn More
Raybiotech Inc Human/Mouse/Rat Phospho-Raf-1 (Ser301) and Total Raf-1 ELISA Kit.
Supplier:  Raybiotech Inc PELRAF1S301T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated Raf-1 (Ser301) and Total Raf-1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human, mouse, and rat Raf-1 phosphorylated at site Serine-301 and Total Raf-1.
Catalog No. 50-195-0704