Learn More
Raybiotech Inc Human Phospho-RSK1 (S380) ELISA and Total RSK1 ELISA Kit.
Supplier:  Raybiotech Inc PELRSK1S380T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated RSK1 (Ser380) and Total RSK1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes Human RSK1 phosphorylated at site Serine-380 and Total RSK1.
Catalog No. 50-195-0717