Learn More
Raybiotech Inc Human, Mouse, Rat Phospho-SHC (Tyr427) and Total SHC ELISA Kit.
Supplier:  Raybiotech Inc PELSHCY427T1
Encompass_Preferred
This assay semi-quantitatively measures SHC phosphorylated at Tyrosine-427 as well as total SHC in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse, Rat SHC phosphorylated at site Tyrosine-427 as well as total SHC.
Catalog No. 50-195-0722