Learn More
Raybiotech Inc Human Phospho-SMAD4 (Thr277) and Total SMAD4 ELISA Kit.
Supplier:  Raybiotech Inc PELSMAD4T277T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated SMAD4 (Thr277) and Total SMAD4 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human SMAD4 phosphorylated at site Threonine-277 as well as total SMAD4.
Catalog No. 50-195-0734