Learn More
Raybiotech Inc Human Phospho-SHIP1 (Tyr1020) and Total SHIP1 ELISA Kit.
Supplier:  Raybiotech Inc PELSHIP1Y1020T1
Encompass_Preferred
This assay semi-quantitatively measures SHIP1 phosphorylated at Tyrosine-1020 as well as total SHIP1 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human SHIP1 phosphorylated at site Tyrosine-1020 as well as total SHIP1.
Catalog No. 50-195-0724