Learn More
Raybiotech Inc Human Phospho-STAT5A (Tyr694) ELISA Kit.
Supplier:  Raybiotech Inc PELSTAT5AY6941
Encompass_Preferred
This assay semi-quantitatively measures STAT5A phosphorylated at Tyrosine-694 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human STAT5A phosphorylated at site Tyrosine-694.
Catalog No. 50-195-0750