Learn More
Raybiotech Inc Human, Mouse, Rat Phospho-TBK1 (Ser172) and Total TBK1 ELISA Kit.
Supplier:  Raybiotech Inc PELTBK1S172T1
Encompass_Preferred
This assay semi-quantitatively measures TBK1 phosphorylated at Serine-172 as well as total TBK1 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse, Rat TBK1 phosphorylated at site Serine-172 as well as total TBK1.
Catalog No. 50-195-0760