Learn More
Raybiotech Inc Human Phospho-STAT6 (Tyr641) and Total STAT6 ELISA Kit.
Supplier:  Raybiotech Inc PELSTAT6Y641T1
Encompass_Preferred
This assay semi-quantitatively measures STAT6 phosphorylated at Tyrosine-641 as well as total STAT6 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human STAT6 phosphorylated at site Tyrosine-641 as well as total STAT6.
Catalog No. 50-195-0755