Learn More
Raybiotech Inc Human Phosphotyrosine TrkB ELISA Kit.
Supplier:  Raybiotech Inc PELTRKBY1
Encompass_Preferred
This assay semi-quantitatively measures tyrosine-phosphorylated TrkB in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes human TrkB phosphotyrosine.
Catalog No. 50-195-0764