Learn More
Raybiotech Inc Human, Mouse Phospho-ULK1 (S556) and Total ULK1 ELISA Kit.
Supplier:  Raybiotech Inc PELULK1S556T1
Encompass_Preferred
This assay semi-quantitatively measures ULK1 phosphorylated at Serine-556 as well as total ULK1 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse ULK1 phosphorylated at site Serine-556 as well as total ULK1.
Catalog No. 50-195-0770