Learn More
Raybiotech Inc Human Phospho-WNK1 (T60) & Total WNK1 ELISA Kit.
Supplier:  Raybiotech Inc PELWNK1T60T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated WNK1 (Thr60) & Total WNK1 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes human WNK1 phosphorylated at site Threonine-60 and total WNK1.
Catalog No. 50-195-0776