Learn More
Raybiotech Inc Human Phosphotyrosine VEGFR2 ELISA Kit.
Supplier:  Raybiotech Inc PELVEGFR2Y1
Encompass_Preferred
This assay semi-quantitatively measures phosphotyrosine VEGFR2 in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes human SMAD4 phosphotyrosine.
Catalog No. 50-195-0771