Learn More
Raybiotech Inc Human c-Fos Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHCFOS1
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical c-Fos binding sequences to semi-quantitatively detect active c-Fos in lysates or nuclear extracts. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0913