Learn More
Raybiotech Inc Human ELK-1 Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHELK11
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical ELK-1 binding sequences to semi-quantitatively detect active ELK-1 in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0916