Learn More
Raybiotech Inc Human Fra1 Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHFRA11
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical Fra1 binding sequences to semi-quantitatively detect active Fra1 in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0919