Learn More
Raybiotech Inc Human GATA-3 Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHGATA31
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical GATA3 binding sequences to semi-quantitatively detect active GATA3 in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0923