Learn More
Raybiotech Inc Human HIF-1α Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHHIF1A1
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical HIF-1α binding sequences to semi-quantitatively detect active HIF-1α in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0924